Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AMOTL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120481
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120481 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120481 Supplier Invitrogen™ Supplier No. PA5120481
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: 293T, U-251MG, HepG2, Mouse liver, Mouse brain. Immunogen sequence: RQFAMDAAAT AAAQRDTTLI RHSPQPSPSS SFNEGLLTGG HRHQEMESRL KVLHAQILEK DAVIKVLQQR SRRDPGKAIQ GSLRPAKSVP SVFAAAAAGT QGWQGLSSSE RQTADAPARL TTDRAPTEEP VVTAPPAAHA KHGSRDGSTQ TEGPPDSTST CLPPEPDSLL GCSSSQRAAS LDSVATSRVQ DLSDMVEILI.

Regulates the translocation of phosphorylated SRC to peripheral cell-matrix adhesion sites. Required for proper architecture of actin filaments. Inhibits the Wnt/beta-catenin signaling pathway, probably by recruiting CTNNB1 to recycling endosomes and hence preventing its translocation to the nucleus. Participates in angiogenesis. May play a role in the polarity, proliferation and migration of endothelial cells. Selectively promotes FGF-induced MAPK activation through SRC.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AMOTL2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.14 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene AMOTL2
Gene Accession No. Q8K371, Q9Y2J4
Gene Alias AMOTL2; angiomotin like 2; angiomotin-like 2; angiomotin-like protein 2; AW549739; C79691; KIAA0989; LCCP; Leman coiled-coil protein; MASCOT; mKIAA0989
Gene Symbols AMOTL2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 638-837 of human AMOTL2 (NP_001265612.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51421, 56332
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.