Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amoxicillin sodium salt, 87% min.

Catalog No. p-7057531
Click to view available options
Quantity:
5 g
25 g

Catalog No. AAJ6667506

Supplier: Thermo Scientific Chemicals J6667506

Only null left
Add to Cart

CAS: 34642-77-8 | C16H18N3NaO5S | 387.386 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34642-77-8
Molecular Formula C16H18N3NaO5S
Molecular Weight (g/mol) 387.386
MDL Number MFCD08063910
InChI Key BYHDFCISJXIVBV-ZONSQSGVSA-M
PubChem CID 131632470
IUPAC Name sodium;(2S,5R,6S)-6-[[2-amino-2-(4-hydroxyphenyl)acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
SMILES CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=C(C=C3)O)N)C(=O)[O-])C.[Na+]

Specifications

Type Amoxicillin Sodium Salt
Physical Form Powder
Melting Point 130°C
Color White
Quantity 5 g
Flash Point >110°C (230°F)
Boiling Point 743°C
Solubility Information Soluble in water.
Formula Weight 387.39
Percent Purity ≥89%
Chemical Name or Material Amoxicillin sodium salt
Show More Show Less
Hazard Category H317-H334
Hazard Statement GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Precautionary Statement P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
EINECSNumber 252-124-1
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.