Learn More
Ampco Safety Tools Tool Kits
SDP
Catalog No. 19805934 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Product Type:
11-piece Tool Kit
17-piece Hazmat Tool Kit
4-piece Tool Kit
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19-805-934 11-piece Tool Kit
19-805-933 4-piece Tool Kit
19-805-935 17-piece Hazmat Tool Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-805-934 Supplier Ampco Safety Tools Supplier No. M48
Add to Cart
Add to Cart
Includes
Four-piece tool kit
(19-805-933) includes: standard screwdriver (5/16 in. tip, 3 in. blade), 8 in. adjustable end wrench, 9 in. crate opener, 6.50 in. combination pliers.
11-piece tool kit
(19-805-934) includes: two standard screwdrivers (5/16 in. tip, 3 in. blade and5/16 in. tip, 6 in. blade), three adjustable end wrenches (6, 8, and 10 in.), 9 in. crate opener, No. 10 groove joint pliers, two No. 2 Phillips screwdriver, 10 in. adjustable pipe wrench, 16 oz. claw hammer with fiberglass handle.
16-piece tool kit
(19-120-3787) includes: four double end box wrenches (5/16 x 3/8 in., 7/16 x 1/2 in., 9/16 x 5/8 in., 3/4 x 7/8 in.), No. 2 Phillips screwdriver, standard screwdriver (5/16 in. tip, 6 in. blade), 10 in. adjustable wrench, two combination pliers (61/2 and 8 in.), 10 in. groove joint pliers, common knife (31/8 in. blade, 63/4 in. OAL), stiff putty knife (11/4 in.
Nonsparking tools with Factory Mutual approval
  • Specially engineered for use in safety-sensitive applications to reduce the risk of fire and explosions
  • Nonsparking, nonmagnetic, and corrosion resistant
  • Made in the USA
Certifications
FM approved.
Compliance
Designed using Federal, military, ANSI™, and DIN standards.
Specifications
Product Type 11-piece Tool Kit
For Use With (Equipment) Safety-sensitive applications to reduce the risk of fire and explosions
Includes 8 in. Adjustable combination pliers, shoe handle brush, K-2 putty knife, 1.25 x 3.56 in. blade, 7.125 in. OAL, K-1 common knife, 3.125 in. blade, 6.75 in. OAL, 10 in. groove joint pliers, No.2 Phillips screwdriver, S-49 standard screwdriver, 5.166 in. tip, 6 in. blade, 16 oz. claw hammer with Fiberglass handle, 6 in. adjustable end wrench, 10 in. adjustable end wrench, 10 in. adjustable pipe wrench, tool box
Weight (English) 10.2 lb.
Weight (Metric) 4.6 kg