Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AMPD1 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP224509MFV610
Description
AMPD1 Polyclonal antibody specifically detects AMPD1 in Human, Mouse, Rat, Bovine, Canine, Opossum, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
AMPD1 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
adenosine monophosphate deaminase 1, adenosine monophosphate deaminase 1 (isoform M), adenosine monophosphate deaminase-1 (muscle), AMP deaminase 1, AMP deaminase isoform M, AMPD, EC 3.5.4.6, MAD, MADA, Myoadenylate deaminase, skeletal muscle AMPD | |
A portion of amino acids 140-190 of human AMPDA1 was used as the immunogen. | |
0.1 mL | |
Cancer, Endocrinology, Signal Transduction | |
270 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Bovine, Canine | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction