Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AMPD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555761
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555761 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555761 Supplier Invitrogen™ Supplier No. PA555761
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AEEKQIDDAM RNFAEKVFAS EVKDEGGRQE ISPFDVDEIC PISHHEMQAH IFHLETLSTS TEARRKKRFQ GRKTVNLSIP L Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 81%.

AMP deaminase (AMPD) is an allosteric enzyme involved in the regulation of adenosine metabolism. It catalyzes a central reaction in purine nucleotide biosynthesis where AMP is deaminated to IMP, liberating ammonia. There are three functional isoforms of AMPD. AMPD1 (E.C. No 3.5.4.6.) is the skeletal muscle-specific isoform M located in type II muscle fibers, neuromuscular junctions and in capillaries. This protein is the predominant member of AMPD multi-gene family and is considered as the sensor of the cell's changing energy requirements. AMPD1 deficiency causes irregular muscle metabolism due to lower rate of ATP degradation, phosphocreatine hydrolysis and accumulation of lactic acid. Mutated AMPD1 expression has been detected in neuromuscular disorders, exercise-induced skeletal muscle myopathies and congestive heart failures due to coronary artery diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AMPD1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AMPD1
Gene Accession No. P23109
Gene Alias adenosine monophosphate deaminase 1; adenosine monophosphate deaminase 1 (isoform M); adenosine monophosphate deaminase-1 (muscle); AI553520; AMP deaminase 1; AMP deaminase isoform M; Ampd; Ampd01; AMPD1; Ampd-1; MAD; MADA; MMDD; myoadenylate deaminase; RATAMPD01; RP5-1000E10.1; skeletal muscle AMPD
Gene Symbols AMPD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human AMPD1. Recombinant protein control fragment (Product #RP-94088).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 270
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.