Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AMPD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA526127
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA526127 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA526127 Supplier Invitrogen™ Supplier No. PA526127
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AMPD2 Polyclonal Antibody for Western Blot, IHC (P), Flow

Adenosine monophosphate deaminase-2 (EC 3.5.4.6) catalyzes the deamination of AMP to IMP and plays an important role in the purine nucleotide cycle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AMPD2
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Ampd2
Gene Accession No. Q01433
Gene Alias 1200014F01Rik; adenosine monophosphate deaminase 2; adenosine monophosphate deaminase 2 (isoform L); adenosine monophosphate deaminase 2 tv2; AI552571; AMP deaminase 2; AMP deaminase isoform L; AMPD; AMPD2; Ampd-2; PCH9; SPG63
Gene Symbols Ampd2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 187-217 amino acids from the central region of human AMPD2.
Purification Method Ammonium sulfate precipitation, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 271
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.