Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AMPD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545596
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545596 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545596 Supplier Invitrogen™ Supplier No. PA545596
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: VVRALFIREK YMALSLQSFC PTTRRYLQQL AEKPLETRTY EQGPDTPVSA Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 100%; Rat: 93%.

Adenosine monophosphate deaminase-2 (EC 3.5.4.6) catalyzes the deamination of AMP to IMP and plays an important role in the purine nucleotide cycle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AMPD2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Ampd2
Gene Accession No. Q01433
Gene Alias 1200014F01Rik; adenosine monophosphate deaminase 2; adenosine monophosphate deaminase 2 (isoform L); adenosine monophosphate deaminase 2 tv2; AI552571; AMP deaminase 2; AMP deaminase isoform L; AMPD; AMPD2; Ampd-2; PCH9; SPG63
Gene Symbols Ampd2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human AMPD2 (aa 244-293).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 271
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.