Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AMPD3 Antibody (AMPD3/901) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$610.00
Specifications
Antigen | AMPD3 |
---|---|
Clone | AMPD3/901 |
Concentration | 1.0 mg/mL |
Dilution | Flow Cytometry : 1 - 2 ug/million cells in 0.1ml, Immunohistochemistry-Paraffin : 2 - 4 ug/ml, Immunofluorescence : 0.5 - 1.0 ug/ml, Immunocytochemistry : 1 - 2.0 ug/ml, CyTOF-ready |
Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, CyTOF |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP247691A
![]() |
Novus Biologicals
NBP2476910.1MG |
0.1 mg |
Each for $610.00
|
|
|||||
NBP247691B
![]() |
Novus Biologicals
NBP2476910.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
AMPD3 Monoclonal specifically detects AMPD3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
AMPD3 | |
1.0 mg/mL | |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, CyTOF | |
Unconjugated | |
Mouse | |
Human | |
adenosine monophosphate deaminase (isoform E), adenosine monophosphate deaminase 3, AMP aminohydrolase, AMP deaminase 3, AMP deaminase isoform E, EC 3.5.4.6, Erythrocyte AMP deaminase, erythrocyte type AMP deaminase, erythrocyte-specific AMP deaminase, myoadenylate deaminase | |
AMPD3 | |
IgG2b κ | |
Protein A or G purified | |
It recognizes a protein of ∼90kDa, which is identified as Adenosine Monophosphate Deaminase, isoform E (AMPD3). It has 767 amino acids and is assigned an EC 3.5.4.6. It is a highly regulated enzyme that catalyzes the hydrolytic deamination of adenosine monophosphate to inosine monophosphate, a branch point in the adenylate catabolic pathway. AMPD3 gene encodes the erythrocyte (E) isoforms, whereas other family members encode isoforms that predominate in muscle (M) and liver (L) cells. This MAb shows reactivity with cells of the erythroid lineage at all stages of maturation in the peripheral blood, bone marrow, and fetal liver. Non-erythroid lineages are negative by flow cytometry. This MAb is useful in the diagnosis of erythroleukemia, identification of bone marrow erythroid precursors, gating erythroid nucleated precursor cells from malignant cells in bone marrow specimens. |
AMPD3/901 | |
Flow Cytometry : 1 - 2 ug/million cells in 0.1ml, Immunohistochemistry-Paraffin : 2 - 4 ug/ml, Immunofluorescence : 0.5 - 1.0 ug/ml, Immunocytochemistry : 1 - 2.0 ug/ml, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
PBS with No Preservative | |
272 | |
Recombinant full-length human AMDP3 protein | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title