Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AMPD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565218
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565218 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565218 Supplier Invitrogen™ Supplier No. PA565218
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PYCLDDAPPNL DYLVHMQGGI LFVYDNKKML EHQEPHSLPY PDLETYTVDM SHILALITDG PTKTYCHRRL NFLES Highest antigen sequence identity to the following orthologs - mouse 89%, rat 89%.

AMPD3 is a member of the AMP deaminase gene family. This protein is a highly regulated enzyme that catalyzes the hydrolytic deamination of adenosine monophosphate to inosine monophosphate, a branch point in the adenylate catabolic pathway. The protein is the erythrocyte (E) isoforms, whereas other family members isoforms predominate in muscle (M) and liver (L) cells. Mutations in this gene lead to the clinically asymptomatic, autosomal recessive condition erythrocyte AMP deaminase deficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AMPD3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Ampd3
Gene Accession No. Q01432
Gene Alias adenosine monophosphate deaminase (isoform E); adenosine monophosphate deaminase 3; AMP aminohydrolase; AMP deaminase 3; AMP deaminase H-type; AMP deaminase isoform E; Ampd; Ampd3; Erythrocyte AMP deaminase; erythrocyte type AMP deaminase; erythrocyte-specific AMP deaminase; heart-type AMPD; myoadenylate deaminase
Gene Symbols Ampd3
Host Species Rabbit
Immunogen Recombinant Human AMPD3. Recombinant protein control fragment (Product #RP-104691).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 272
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.