Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AMPD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576912
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA576912 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA576912 Supplier Invitrogen™ Supplier No. PA576912
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is >95% (by SDS-PAGE).

AMPD3 is a member of the AMP deaminase gene family. This protein is a highly regulated enzyme that catalyzes the hydrolytic deamination of adenosine monophosphate to inosine monophosphate, a branch point in the adenylate catabolic pathway. The protein is the erythrocyte (E) isoforms, whereas other family members isoforms predominate in muscle (M) and liver (L) cells. Mutations in this gene lead to the clinically asymptomatic, autosomal recessive condition erythrocyte AMP deaminase deficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AMPD3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene Ampd3
Gene Accession No. Q01432
Gene Alias adenosine monophosphate deaminase (isoform E); adenosine monophosphate deaminase 3; AMP aminohydrolase; AMP deaminase 3; AMP deaminase H-type; AMP deaminase isoform E; Ampd; Ampd3; Erythrocyte AMP deaminase; erythrocyte type AMP deaminase; erythrocyte-specific AMP deaminase; heart-type AMPD; myoadenylate deaminase
Gene Symbols Ampd3
Host Species Rabbit
Immunogen Recombinant full length Human AMPD3.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 272
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.