Learn More
Invitrogen™ AMPD3 Polyclonal Antibody

Description
AMPD3 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | AMPD3 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | Ampd3 |
| Gene Accession No. | O08739, Q01432 |
| Gene Alias | adenosine monophosphate deaminase (isoform E); adenosine monophosphate deaminase 3; AMP aminohydrolase; AMP deaminase 3; AMP deaminase H-type; AMP deaminase isoform E; Ampd; Ampd3; Erythrocyte AMP deaminase; erythrocyte type AMP deaminase; erythrocyte-specific AMP deaminase; heart-type AMPD; myoadenylate deaminase |
| Gene Symbols | Ampd3 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.