Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AMPD3 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP304594100UL
Description
AMPD3 Polyclonal specifically detects AMPD3 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
AMPD3 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
272 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Immunofluorescence | |
Unconjugated | |
adenosine monophosphate deaminase (isoform E), adenosine monophosphate deaminase 3, AMP aminohydrolase, AMP deaminase 3, AMP deaminase isoform E, EC 3.5.4.6, Erythrocyte AMP deaminase, erythrocyte type AMP deaminase, erythrocyte-specific AMP deaminase, myoadenylate deaminase | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human AMPD3 (NP_000471.1). | |
100 μL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction