Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(±)-Amphetamine-d6 solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant

Catalog No. A0451ML Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 mL

Catalog No. A0451ML

Supplier: MilliporeSigma A0451ML

Only null left
Add to Cart
Add to Cart

Amphetamine, a stimulant of the phenethylamine class, is found in ADHD-treatment drugs.

Amphetamine, a stimulant of the phenethylamine class, is found in ADHD-treatment drugs such as Adderall™, Vyvanse™, and ProCentra™. Because of its stimulant effect, amphetamine is used recreationally as a performance-enhancer and illicit drug. This stable-labeled internal standard is suitable for quantitation of amphetamine levels in urine, serum, or plasma by LC/MS or GC/MS for urine drug testing, clinical toxicology, forensic analysis, or sports testing.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (±)-Amphetamine-d6
Packaging Ampule of 1 mL
CAS 73758-26-6
Molecular Formula C9D6H7N
Concentration 1.0 mg/mL in Methanol
For Use With (Equipment) Gas Chromatography (GC), Liquid Chromatography (LC)
Solution Type Standard Solution
Type Certified Reference Material
Industry Type Forensics, Toxicology
Molecular Weight (g/mol) 141.24
Quantity 1 mL
Grade Certified
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.