Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Amphiregulin Antibody (RM0151-7D17), mFluor Violet 610 SE, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP212036MFV610
Description
Amphiregulin Monoclonal antibody specifically detects Amphiregulin in Mouse samples. It is validated for Western BlotSpecifications
Amphiregulin | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Western Blot | |
RM0151-7D17 | |
Western Blot | |
amphiregulin, AR, AREG, AREGB, Colorectum cell-derived growth factor, CRDGF, MGC13647, schwannoma-derived growth factor, SDGF | |
Mouse recombinant Amphiregulin | |
0.1 mL | |
Biologically Active Proteins, Cell Biology, Cytokine Research, Growth and Development, Signal Transduction | |
374 | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction