Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Amphiregulin Monoclonal Antibody (A5)

Mouse Monoclonal Antibody
$194.00 - $516.00
Specifications
| Antigen | Amphiregulin |
|---|---|
| Clone | A5 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA541548
![]() |
Invitrogen™
MA541548 |
200 μL |
Each for $516.00
|
|
|||||
|
PIMA543967
|
Invitrogen™
MA543967 |
20 μL |
Each for $194.00
|
|
|||||
Description
Amphiregulin Monoclonal Antibody for Western Blot, IHC (P)
Amphiregulin (AR) binds to EGF-Receptor (EGFR) with lower affinity than EGF. The mature secreted form AR is an 84-amino acid residue glycosylated polypeptide growth regulator, which is generated by proteolytic processing of a 252-amino acid transmembrane precursor. Seven different polypeptide ligands, which derive from distinct genes, are capable of binding to the extracellular domain of EGFR. These ligands include EGF, TGFalpha, AR, HB-EGF, cripto-1, epiregulin, and betacellulin. All of these growth factors contain a characteristic EGF-like domain which is defined by 6 evenly spaced cysteine residues that generate 3 loops through the formation of disulfide bonds. AR protein has been localized by immunohistochemistry to the epithelium of the colon, stomach, pancreas, breast, and placenta. AR is reportedly overexpressed in human cancers of breast, colon, stomach, and pancreas.Specifications
| Amphiregulin | |
| 1 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P15514 | |
| 374 | |
| Recombinant protein Amphiregulin. The antigen corresponds to amino acid range 20-100 of the target protein. | |
| Antibody |
| A5 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Monoclonal | |
| Liquid | |
| IgG2b κ | |
| Human | |
| AREG | |
| amphiregulin; amphiregulin B; AR; AREG; AREGB; colorectum cell-derived growth factor; CRDGF; Mcub; MGC13647; schwannoma-derived growth factor; SDGF | |
| AREG | |
| Primary | |
| Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title