Learn More
Promega AMPK (A1/B1/G1) Kinase Enzyme System, 1mg
Catalog No. PRV1922 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV1922 1 mg
1 options
Catalog No. PRV1922 Supplier Promega Supplier No. V1922
Add to Cart
Add to Cart
Request a Quote
Recombinant full-length human AMPK (combination of A1/B1/G1 subunits) was expressed by baculovirus in Sf9 insect cells using C-terminal His tags. AMPK is an important energy-sensing enzyme that monitors cellular energy status.
  • Easily Screen and Profile AMPK (A1/B1/G1) Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C