Learn More
Promega AMPK (A1/B1/G2) Kinase Enzyme System, 10microg
Catalog No. PRV4012 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4012 10 μg
1 options
Catalog No. PRV4012 Supplier Promega Supplier No. V4012
Recombinant full-length human AMPK (combination of A1/B1/G2 subunits) was expressed by baculovirus in Sf9 insect cells using a C-terminal His tag. AMP-activated protein kinase (AMPK) exhibits a key role as master regulator of cellular energy homeostasis.
  • Easily Screen and Profile AMPK (A1/B1/G2) Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C