Learn More
Promega AMPK (A1/B1/G2) Kinase Enzyme System, 1mg
Catalog No. PRV6274 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6274 1 mg
1 options
Catalog No. PRV6274 Supplier Promega Supplier No. V6274
Add to Cart
Add to Cart
Request a Quote
Recombinant full-length human AMPK (combination of A1/B1/G2 subunits) was expressed by baculovirus in Sf9 insect cells using a C-terminal His tag. AMP-activated protein kinase (AMPK) exhibits a key role as master regulator of cellular energy homeostasis.
  • Easily Screen and Profile AMPK (A1/B1/G2) Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for convenient bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C