Learn More
Promega AMPK (A2/B1/G1) Kinase Enzyme System, 10microg
Catalog No. PRV4014 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4014 10 μg
1 options
Catalog No. PRV4014 Supplier Promega Supplier No. V4014
Recombinant full-length human AMPK (A2/B1/G1 subunits) was expressed by baculovirus in Sf9 insect cells using a C-terminal His tag. AMPK (A2/B1/G1) plays a key role in insulin signaling pathway and is a major therapeutic target for diabetes treatment.
  • Easily Screen and Profile AMPK (A2/B1/G1) Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C