Learn More
Promega AMPK (A2/B1/G1) Kinase Enzyme System, 1mg
Catalog No. PRV6275 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6275 1 mg
1 options
Catalog No. PRV6275 Supplier Promega Supplier No. V6275
Add to Cart
Add to Cart
Request a Quote
Recombinant full-length human AMPK (A2/B1/G1 subunits) was expressed by baculovirus in Sf9 insect cells using a C-terminal His tag. AMPK (A2/B1/G1) plays a key role in insulin signaling pathway and is a major therapeutic target for diabetes treatment.
  • Easily Screen and Profile AMPK (A2/B1/G1) Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C