Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMPK alpha 1 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501571306
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-1306 10 μL
50-157-1305 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-157-1306 Supplier Bethyl Laboratories, Inc Supplier No. IHC00002T
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat, Chicken, Turkey, Bovine, Dog, Horse, Rabbit, Guinea pig_10141, Pig, Panda, Orangutan,

AMPKa belongs to the family of AMP activated protein kinases, that plays a central role in regulating cellular and organismal energy balance in response to the balance between AMP/ATP, and intracellular Ca2+ levels. AMPK exists as a heterotrimer comprising of catalytic alpha subunit, and regulatory beta and gamma subunits. The alpha subunit is activated by increases in the cellular AMP:ATP ratio caused by metabolic stresses, and can mediate the effects of the adipokines leptin and adiponectin. Two isoforms differ in subcellular localization and AMP-dependence. Phosphorylation of threonine 172 in the alpha subunit is a key determit of AMPK activity.

Specifications

Antigen AMPK alpha 1
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene PRKAA1
Gene Accession No. Q13131, Q5EG47
Gene Alias AMPK, AMPKa1, MGC33776, MGC57364
Gene Symbols PRKAA1
Host Species Rabbit
Immunogen Between 350 and 400
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 105787, 5562
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.