Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AMPK alpha 2, p Ser491 Antibody (3H10), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32640150UL
Description
AMPK alpha 2 Monoclonal antibody specifically detects AMPK alpha 2 in Human samples. It is validated for Western Blot, ELISASpecifications
AMPK alpha 2 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA | |
3H10 | |
Western Blot 1:500-1:5000, ELISA | |
5'-AMP-activated protein kinase catalytic subunit alpha-2, AMPK subunit alpha-2, AMPK2, AMPK5'-AMP-activated protein kinase, catalytic alpha-2 chain, AMPK-alpha-2 chain, EC 2.7.11, EC 2.7.11.1, PRKAA, protein kinase, AMP-activated, alpha 2 catalytic subunit | |
A synthesized peptide derived from Human AMPK alpha 2 [p Ser491] [UniProt P54646] | |
50 μL | |
Autophagy, Cancer, Hypoxia, MAP Kinase Signaling, mTOR Pathway, Phospho Specific, Protein Kinase, Signal Transduction, Translation Control | |
5563 | |
Store at -20 to -70C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction