Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AMPK alpha 2, p Ser491 Antibody (SR2078), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP321566100UL
Description
AMPK alpha 2 Monoclonal antibody specifically detects AMPK alpha 2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
AMPK alpha 2 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
SR2078 | |
Western Blot 1:1000-1:2000 | |
5'-AMP-activated protein kinase catalytic subunit alpha-2, AMPK subunit alpha-2, AMPK2, AMPK5'-AMP-activated protein kinase, catalytic alpha-2 chain, AMPK-alpha-2 chain, EC 2.7.11, EC 2.7.11.1, PRKAA, protein kinase, AMP-activated, alpha 2 catalytic subunit | |
A synthesized peptide derived from human phospho-AMPK alpha 2 [Ser491] (Uniprot #: P54646) | |
100 μg | |
Autophagy, Cancer, Hypoxia, MAP Kinase Signaling, mTOR Pathway, Phospho Specific, Protein Kinase, Signal Transduction, Translation Control | |
5563 | |
Store at -20 degrees C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction