Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMPK alpha 2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984955
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4955 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4955 Supplier Bioss Supplier No. BS2771R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene is a catalytic subunit of the AMP-activated protein kinase (AMPK). AMPK is a heterotrimer consisting of an alpha catalytic subunit, and non-catalytic beta and gamma subunits. AMPK is an important energy-sensing enzyme that monitors cellular energy status. In response to cellular metabolic stresses, AMPK is activated, and thus phosphorylates and inactivates acetyl-CoA carboxylase (ACC) and beta-hydroxy beta-methylglutaryl-CoA reductase (HMGCR), key enzymes involved in regulating de novo biosynthesis of fatty acid and cholesterol. Studies of the mouse counterpart suggest that this catalytic subunit may control whole-body insulin sensitivity and is necessary for maintaining myocardial energy homeostasis during ischemia.

Specifications

Antigen AMPK alpha 2
Applications ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PRKAA2
Gene Accession No. P54646
Gene Alias 2310008I11Rik; 5′-AMP-activated protein kinase catalytic subunit alpha-2; 5′-AMP-activated protein kinase, catalytic alpha-2 chain; A830082D05; ACACA kinase; Acetyl-CoA carboxylase kinase; AMP-activated protein kinase; AMP-activated protein kinase alpha-2 subunit variant 2; AMP-activated protein kinase alpha-2 subunit variant 3; AMP-activated protein kinase alpha-2 variant B; AMPK; AMPK alpha 2; AMPK alpha-2 chain; AMPK subunit alpha-2; AMPK2; AMPKa2; AMPKalpha2; AMPK-alpha-2 chain; EC 2.7.11.1; HMGCR kinase; Hydroxymethylglutaryl-CoA reductase kinase; PRKAA; Prkaa2; protein kinase AMP-activated catalytic subunit alpha 2; protein kinase, AMP-activated, alpha 2 catalytic subunit
Gene Symbols PRKAA2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human AMPK alpha 2 .
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5563
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.