Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMPK beta 1 + 2 Rabbit anti-Human∣Mouse∣Rat, Polyclonal, Bioss
SDP

Catalog No. 501981137
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1137 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1137 Supplier Bioss Supplier No. BS1116R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen AMPK beta 1 + 2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Prkab1
Gene Accession No. O43741, P80386, Q6PAM0, Q9QZH4, Q9R078, Q9Y478
Gene Alias 1300015D22Rik; 5730553K21Rik; 5′-AMP-activated protein kinase 40 kDa subunit; 5-AMP-activated protein kinase beta subunit; 5′-AMP-activated protein kinase beta-1 subunit; 5′-AMP-activated protein kinase subunit beta-1; 5′-AMP-activated protein kinase subunit beta-1-like protein; 5′-AMP-activated protein kinase subunit beta-2; 5′-AMP-activated protein kinase, beta subunit; 5′-AMP-activated protein kinase, beta-2 subunit; AMP-activated protein kinase beta 2 non-catalytic subunit; AMP-activated protein kinase beta subunit; AMP-activated protein kinase beta-2 regulatory subunit; AMPK; AMPK beta 1; AMPK beta -1 chain; AMPK beta 2; AMPK beta-1 chain; AMPK beta-2 chain; AMPK subunit beta-1; AMPK subunit beta-2; AMPKb; AU021155; AW049591; BB124140; E430008F22; HAMPKb; I79_012592; Prkab1; PRKAB2; protein kinase AMP-activated non-catalytic subunit beta 1; protein kinase AMP-activated non-catalytic subunit beta 2; protein kinase, AMP-activated, beta 1 non-catalytic subunit; protein kinase, AMP-activated, beta 2 non-catalytic subunit; protein kinase, AMP-activated, noncatalytic, beta-1
Gene Symbols Prkab1, PRKAB2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human AMPK beta 1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108097, 19079, 5564, 5565, 64562, 83803
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.