Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AMPK beta 2/PRKAB2 Antibody (OTI4H4), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271226G
Description
AMPK beta 2/PRKAB2 Monoclonal specifically detects AMPK beta 2/PRKAB2 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
AMPK beta 2/PRKAB2 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PRKAB2 | |
Full length recombinant protein of human AMPK beta 2/PRKAB2 (NP_005390) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot | |
OTI4H4 | |
Western Blot | |
5′-AMP-activated protein kinase, beta-2 subunit, AMPK beta 2,5′-AMP-activated protein kinase subunit beta-2, AMPK beta-2 chain, AMPK subunit beta-2, MGC61468, protein kinase, AMP-activated, beta 2 non-catalytic subunit | |
Mouse | |
Affinity Purified | |
Lipid and Metabolism | |
5565.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction