Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMPK gamma 1/2/3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989073
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9073 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9073 Supplier Bioss Supplier No. BS9446R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The 5-AMP activated protein kinase (AMPK), a member of the SNF1 (sucrose non fermentor) kinase family, is a heterotrimeric protein comprise of alpha (63 kDa), beta (30 kDa) and gamma (38 kDa) subunits. The alpha subunit is the catalytic subunit, while beta and gamma are non catalytic subunits (although they have been found to interact with the active subunit in liver). AMPK regulates fatty acid and sterol synthesis by phosphorylation of acetyl-CoA as well as cholesterol synthesis via phosphorylation and inactivation of hydroxymethylglutaryl-CoA reductase. AMPK is activated by AMP and can be also regulated by treatment with purified protein phosphatase in vitro.

Specifications

Antigen AMPK gamma 1/2/3
Applications ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PRKAG2
Gene Accession No. P54619, Q9UGI9, Q9UGJ0
Gene Alias 2410051C13Rik; 5′-AMP-activated protein kinase subunit gamma-1; 5′-AMP-activated protein kinase subunit gamma-2; 5′-AMP-activated protein kinase subunit gamma-3; 5′-AMP-activated protein kinase, gamma-1 subunit; 5-AMP-activated protein kinase, gamma-2 subunit; 5′-AMP-activated protein kinase, gamma-3 subunit; AA571379; AAKG; AAKG2; adenosine monophosphate-activated protein kinase gamma 2-subunit; AI854673; Amkg3; AMP-activated protein kinase gamma 3 subunit; AMP-activated protein kinase gamma subunit; AMP-activated protein kinase gamma2 subunit; AMP-activated protein kinase, noncatalytic gamma-1 subunit; AMPK gamma 2; AMPK gamma1; AMPK- gamma1; AMPK gamma-1 chain; AMPK gamma2; AMPK- gamma2; AMPK gamma-2 chain; AMPK gamma3; AMPK- gamma3; AMPK gamma-3 chain; AMPK subunit gamma-1; AMPK subunit gamma-2; AMPK subunit gamma-3; AMPKG; AMPKG3; AMPKg3L; AMPKg3S; BB036179; CMH6; H91620p; Prkaac; Prkag1; PRKAG2; Prkag3; Prkga1; Protein kinase AMP-activated gamma; protein kinase AMP-activated non-catalytic subunit gamma 1; protein kinase AMP-activated non-catalytic subunit gamma 2; protein kinase AMP-activated non-catalytic subunit gamma 3; protein kinase, AMP-activated, gamma 1 non-catalytic subunit; protein kinase, AMP-activated, gamma 2 non-catalytic subunit; protein kinase, AMP-activated, gamma 3 non-catalytic subunit; protein kinase, AMP-activated, gamma 3 non-catatlytic subunit; WPWS
Gene Symbols PRKAG1, PRKAG2, PRKAG3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human PRKAG2/AMPK_2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51422, 53632, 5571
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.