Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AMPK gamma-1 Monoclonal Antibody (7H3E9)
GREENER_CHOICE

Catalog No. PIMA531866
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531866 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531866 Supplier Invitrogen™ Supplier No. MA531866
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31866 has been tested in indirect ELISA.

The protein encoded by this gene is a regulatory subunit of the AMP-activated protein kinase (AMPK). AMPK is a heterotrimer consisting of an alpha catalytic subunit, and non-catalytic beta and gamma subunits. AMPK is an important energy-sensing enzyme that monitors cellular energy status. In response to cellular metabolic stresses, AMPK is activated, and thus phosphorylates and inactivates acetyl-CoA carboxylase (ACC) and beta-hydroxy beta-methylglutaryl-CoA reductase (HMGCR), key enzymes involved in regulating de novo biosynthesis of fatty acid and cholesterol. This subunit is one of the gamma regulatory subunits of AMPK. Alternatively spliced transcript variants encoding distinct isoforms have been observed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AMPK gamma-1
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 7H3E9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene PRKAG1
Gene Accession No. P54619
Gene Alias 5'-AMP-activated protein kinase subunit gamma-1; 5'-AMP-activated protein kinase, gamma-1 subunit; AA571379; AMP-activated protein kinase, noncatalytic gamma-1 subunit; ampk gamma; AMPK gamma1; AMPK gamma-1 chain; AMPK subunit gamma-1; AMPKG; BB036179; Prkaac; PRKAG1; Prkga1; Protein kinase AMP-activated gamma; protein kinase AMP-activated non-catalytic subunit gamma 1; protein kinase, AMP-activated, gamma 1 non-catalytic subunit
Gene Symbols PRKAG1
Host Species Mouse
Immunogen Purified recombinant fragment of human PRKAG1 (AA: 230-331) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5571
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.