Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AMPK gamma-1,2,3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5101131
Description
Antibody detects endogenous levels of total PRKAG1/2/3.
The 5-AMP activated protein kinase (AMPK), a member of the SNF1 (sucrose non fermentor) kinase family, is a heterotrimeric protein comprise of alpha (63 kDa), beta (30 kDa) and gamma (38 kDa) subunits. The alpha subunit is the catalytic subunit, while beta and gamma are non catalytic subunits (although they have been found to interact with the active subunit in liver). AMPK regulates fatty acid and sterol synthesis by phosphorylation of acetyl-CoA as well as cholesterol synthesis via phosphorylation and inactivation of hydroxymethylglutaryl-CoA reductase. AMPK is activated by AMP and can be also regulated by treatment with purified protein phosphatase in vitro.
Specifications
AMPK gamma-1,2,3 | |
Polyclonal | |
Unconjugated | |
PRKAG1 | |
2410051C13Rik; 5'-AMP-activated protein kinase subunit gamma-1; 5'-AMP-activated protein kinase subunit gamma-2; 5'-AMP-activated protein kinase subunit gamma-3; 5'-AMP-activated protein kinase, gamma-1 subunit; 5-AMP-activated protein kinase, gamma-2 subunit; 5'-AMP-activated protein kinase, gamma-3 subunit; AA571379; AAKG; AAKG2; adenosine monophosphate-activated protein kinase gamma 2-subunit; AI854673; Amkg3; AMP-activated protein kinase gamma 3 subunit; AMP-activated protein kinase gamma subunit; AMP-activated protein kinase gamma2 subunit; AMP-activated protein kinase, noncatalytic gamma-1 subunit; ampk gamma; AMPK gamma 2; AMPK gamma1; AMPK- gamma1; AMPK gamma-1 chain; AMPK gamma2; AMPK- gamma2; AMPK gamma-2 chain; AMPK gamma3; AMPK- gamma3; AMPK gamma-3 chain; AMPK subunit gamma-1; AMPK subunit gamma-2; AMPK subunit gamma-3; AMPKg; Ampkg3; AMPKg3L; AMPKg3S; BB036179; CMH6; H91620p; Prkaac; Prkag1; PRKAG2; PRKAG3; Prkga1; Protein kinase AMP-activated gamma; protein kinase AMP-activated non-catalytic subunit gamma 1; protein kinase AMP-activated non-catalytic subunit gamma 2; protein kinase AMP-activated non-catalytic subunit gamma 3; protein kinase, AMP-activated, gamma 1 non-catalytic subunit; protein kinase, AMP-activated, gamma 2 non-catalytic subunit; protein kinase, AMP-activated, gamma 3 non-catalytic subunit; protein kinase, AMP-activated, gamma 3 non-catatlytic subunit; WPWS | |
Rabbit | |
Affinity Chromatography | |
RUO | |
108099, 19082, 241113, 25520, 301518, 373545, 51422, 53632, 5571 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O54950, P54619, P80385, Q8BGM7, Q91WG5, Q9UGI9, Q9UGJ0 | |
PRKAG1, PRKAG2, PRKAG3 | |
A synthesized peptide derived from human PRKAG1(Accession P54619), corresponding to amino acid residues K58-E108. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction