Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AMPK gamma-2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 384200
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-420-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 38-420-0 Supplier Invitrogen™ Supplier No. 384200
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AMPK gamma-2 Polyclonal Antibody for Western Blot

AMP-activated protein kinase (AMPK) is a heterotrimeric protein composed of a catalytic alpha subunit, a noncatalytic beta subunit, and a noncatalytic regulatory gamma subunit. Various forms of each of these subunits exist, encoded by different genes. AMPK is an important energy-sensing enzyme that monitors cellular energy status and functions by inactivating key enzymes involved in regulating de novo biosynthesis of fatty acid and cholesterol. This gene is a member of the AMPK gamma subunit family and encodes a protein with four cystathionine beta-synthase domains. Mutations in this gene have been associated with ventricular pre-excitation (Wolff-Parkinson-White syndrome), progressive conduction system disease and cardiac hypertrophy. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AMPK gamma-2
Applications Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene PRKAG2
Gene Accession No. Q91WG5, Q9UGJ0
Gene Alias 2410051C13Rik; 5'-AMP-activated protein kinase subunit gamma-2; 5-AMP-activated protein kinase, gamma-2 subunit; AAKG; AAKG2; adenosine monophosphate-activated protein kinase gamma 2-subunit; AI854673; AMP-activated protein kinase gamma2 subunit; ampk gamma; AMPK gamma 2; AMPK gamma2; AMPK gamma-2 chain; AMPK subunit gamma-2; CMH6; H91620p; OTTHUMP00000211956; PRKAG2; protein kinase AMP-activated non-catalytic subunit gamma 2; protein kinase, AMP-activated, gamma 2 non-catalytic subunit; WPWS
Gene Symbols PRKAG2
Host Species Rabbit
Immunogen Synthetic peptide derived from the N-terminal region of the human AMPK-gamma-2 (5'-AMP-activated protein kinase, gamma-2subunit, PRKAG2) protein, which differs from mouse by one amino acid.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108099, 51422
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.