Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AMPK gamma-3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113447
Description
AMPK gamma-3 Polyclonal Antibody for Western Blot, ELISA
The protein encoded by this gene is a regulatory subunit of the AMP-activated protein kinase (AMPK). AMPK is a heterotrimer consisting of an alpha catalytic subunit, and non-catalytic beta and gamma subunits. AMPK is an important energy-sensing enzyme that monitors cellular energy status. In response to cellular metabolic stresses, AMPK is activated, and thus phosphorylates and inactivates acetyl-CoA carboxylase (ACC) and beta-hydroxy beta-methylglutaryl-CoA reductase (HMGCR), key enzymes involved in regulating de novo biosynthesis of fatty acid and cholesterol. This subunit is one of the gamma regulatory subunits of AMPK. It is dominantly expressed in skeletal muscle. Studies of the pig counterpart suggest that this subunit may play a key role in the regulation of energy metabolism in skeletal muscle.
Specifications
AMPK gamma-3 | |
Polyclonal | |
Unconjugated | |
PRKAG3 | |
5'-AMP-activated protein kinase subunit gamma-3; 5'-AMP-activated protein kinase, gamma-3 subunit; Amkg3; AMP-activated protein kinase gamma 3 subunit; AMP-activated protein kinase gamma subunit; ampk gamma; AMPK gamma3; AMPK gamma-3 chain; AMPK subunit gamma-3; Ampkg3; AMPKg3L; AMPKg3S; OTTHUMP00000206393; PRKAG3; protein kinase AMP-activated non-catalytic subunit gamma 3; protein kinase, AMP-activated, gamma 3 non-catalytic subunit; protein kinase, AMP-activated, gamma 3 non-catatlytic subunit | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
53632 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
0.49 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9UGI9 | |
PRKAG3 | |
Recombinant Human 5'-AMP-activated protein kinase subunit gamma-3 protein (1-210AA). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction