Description
- Made of polypropylene
- Available with black alpha-numeric
- Flexible, can be cut to desired format
- Autoclavable
- Inside of tube is smooth and has an inert surface
Certifications
RNase, DNase, Pyrogen and DNA-free.
Specifications
Specifications
| Format | Standard |
| Array | 4 x 6 |
| Autoclavable | Autoclavable |
| Color | Yellow |
| Material | Polypropylene |
| No. of Wells | 24 |
| Sterility | Non-sterile |
| No. per Case | 50/Cs. |