Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gilson™ AmpliPurTM Expert Tips

Catalog No. p-7235879 Shop All Gilson Products
Click to view available options
Volume (Metric):
0.5 to 10 μL
10 to 20 μL
10 to 200 μL
100 to 1000 μL

AmpliPurTM Expert Tips are ISO 8655 compliant and provide a significant value for the money spent. These filter tips protect precious samples and your pipettes from cross-contamination and are designed to fit various pipette brands.

  • AmpliPur Expert Tips are designed to fit various pipette brands for maximum convenience on the bench.
  • Equipped with a filter, they also help prevent cross-contamination.
  • Ideal for Molecular Biology.
  • These tips significantly contribute to molecular testing performance by ensuring sample integrity during the pipetting process.
  • Manufactured according to the certified Gilson quality standard, AmpliPur Expert Tips comply with ISO 8655 requirements and are certified free of RNase and DNase, preserving your sample integrity.

Specifications

Volume (Metric) 10 to 20 μL
Filtered Filtered
Sterility Non-sterile
Color Clear
Rack Color Yellow
Autoclavable Not Autoclavable
Length (English) 2 in.
Length (Metric) 52 mm
Certifications/Compliance ISO 8655
Clean Claims Yes
For Use With (Application) Molecular Biology
Compatibility Universal Fit
Format Rack
Tip Style Rectangular Prism
Packaging Tipack
Quantity 960/Pk.
Product Type Air Displacement Tips
Color Code Yellow
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated No
Product Line AmpliPur
Material Polypropylene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.