Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AMT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552166
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552166 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552166 Supplier Invitrogen™ Supplier No. PA552166
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QYRDSHTDSH LHTRQHCSLF DVSHMLQTKI LGSDRVKLME SLVVGDIAEL RPNQGTLSLF TNEAGGILDD LIVTNTSEGH LYVVSNAGCW EKDLALMQDK VRELQNQGRD VGLEVLDNAL L Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 64%.

The enzyme system for cleavage of glycine (glycine cleavage system; EC 2.1.2.10), which is confined to the mitochondria, is composed of 4 protein components: P protein (a pyridoxal phosphate-dependent glycine decarboxylase), H protein (a lipoic acid-containing protein), T protein (a tetrahydrofolate-requiring enzyme), and L protein (a lipoamide dehydrogenase). Glycine encephalopathy (GCE) may be due to a defect in any one of these enzymes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AMT
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AMT
Gene Accession No. P48728
Gene Alias aminomethyltransferase; aminomethyltransferase (glycine cleavage system protein T); aminomethyltransferase, mitochondrial; AMT; EG434437; GCE; GCST; GCVT; glycine cleavage system T protein; NKH
Gene Symbols AMT
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human AMT. Recombinant protein control fragment (Product #RP-88771).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 275
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.