Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AMT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597458
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597458 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597458 Supplier Invitrogen™ Supplier No. PA597458
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AMT Polyclonal Antibody for Western Blot, ELISA

The enzyme system for cleavage of glycine (glycine cleavage system; EC 2.1.2.10), which is confined to the mitochondria, is composed of 4 protein components: P protein (a pyridoxal phosphate-dependent glycine decarboxylase), H protein (a lipoic acid-containing protein), T protein (a tetrahydrofolate-requiring enzyme), and L protein (a lipoamide dehydrogenase). Glycine encephalopathy (GCE) may be due to a defect in any one of these enzymes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AMT
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene AMT
Gene Accession No. P48728, Q8CFA2
Gene Alias aminomethyltransferase; aminomethyltransferase (glycine cleavage system protein T); aminomethyltransferase, mitochondrial; AMT; EG434437; GCE; GCST; GCVT; glycine cleavage system T protein; NKH
Gene Symbols AMT
Host Species Rabbit
Immunogen Recombinant Human Aminomethyltransferase, mitochondrial protein (180-260AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 275, 434437
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.