Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AMT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109385
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109385 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109385 Supplier Invitrogen™ Supplier No. PA5109385
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AELRPNQGTL SLFTNEAGGI LDDLIVTNTS EGHLYVVSNA GCWEKDLALM QDKVRELQNQ GRDVGLEVLD NALLALQGPT AAQVLQAGVA DDLRKLPFMT SAVMEVFGVS GCRVTRCGYT GEDGVEISVP VAGAVHLATA ILKNPEVKLA GLAARDSLRL EAGLCLYGND IDEHTTPVEG SLSWTLGKRR RAAMDFPGAK VIVPQLKGRV QRRRVGLMCE GAPMRAHSPI LNMEGTKIGT VTSGCPSPSL KKNVAMGYVP CEYSRPGTML LVELPSGPCF.

The enzyme system for cleavage of glycine (glycine cleavage system; EC 2.1.2.10), which is confined to the mitochondria, is composed of 4 protein components: P protein (a pyridoxal phosphate-dependent glycine decarboxylase), H protein (a lipoic acid-containing protein), T protein (a tetrahydrofolate-requiring enzyme), and L protein (a lipoamide dehydrogenase). Glycine encephalopathy (GCE) may be due to a defect in any one of these enzymes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AMT
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.72 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene AMT
Gene Accession No. P48728, Q8CFA2
Gene Alias aminomethyltransferase; aminomethyltransferase (glycine cleavage system protein T); aminomethyltransferase, mitochondrial; AMT; EG434437; GCE; GCST; GCVT; glycine cleavage system T protein; NKH
Gene Symbols AMT
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 107-386 of human AMT (NP_001158184.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 275, 306586, 434437
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.