Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AMTN Antibody (OTI2D9), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272507AF405
Description
AMTN Monoclonal specifically detects AMTN in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
AMTN | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
AMTN | |
Full length recombinant protein of human AMTN (NP_997722) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI2D9 | |
Western Blot, Immunohistochemistry | |
amelotin, MGC148132, MGC148133, RSTI689, UNQ689 | |
Mouse | |
Affinity Purified | |
Signal Transduction | |
401138.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction