Learn More
AMTN Mouse anti-Human, Clone: OTI1D6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF808574

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Is a promoter of calcium phosphate mineralization, playing a critical role in the formation of the compact, mineralized, aprismatic enamel surface layer during the maturation stage of amelogenesis.Specifications
AMTN | |
Monoclonal | |
Unconjugated | |
AMTN | |
AI3B, UNQ689 | |
Mouse | |
Affinity Chromatography | |
RUO | |
401138 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI1D6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q6UX39 | |
AMTN | |
Full length human recombit protein of human AMTN produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.