Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMY2A Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985804
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5804 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5804 Supplier Bioss Supplier No. BS3975R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Amylases are secreted proteins that hydrolyze 1,4-alpha-glucoside bonds in oligosaccharides and polysaccharides, and thus catalyze the first step in digestion of dietary starch andglycogen. The human genome has a cluster of several amylase genes that are expressed at high levels in either salivary gland or pancreas.

Specifications

Antigen AMY2A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene AMY2B
Gene Accession No. P04746, P19961
Gene Alias 1,4-alpha-D-glucan glucanohydrolase; 1,4-alpha-D-glucan glucanohydrolase 2B; alpha-amylase; Alpha-amylase 2B; Amy2; Amy2-1; Amy2-2; AMY2A; Amy2a1; Amy2a4; AMY2B; AMY3; amylase; amylase 2-2, pancreatic; amylase 2a1; amylase 2a4; amylase 2a4, pancreatic; amylase 2b; amylase 2b, pancreatic; amylase, alpha 2A (pancreatic); amylase, alpha 2B (pancreatic); Amy-X; Carcinoid alpha-amylase; ENSMUSG00000074267; glycogenase; HXA; mAmy2-1; mAmy2-3; mAmy2-6; OTTMUSG00000022432; OTTMUSG00000022433; OTTMUSG00000022462; PA; Pancreatic alpha-amylase; pancreatic amylase alpha 2A
Gene Symbols AMY2A, AMY2B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human AMY2A.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 279, 280
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.