Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amyl Ether 98.0+%, TCI America™
SDP

Catalog No. A0451500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0451500ML 500 mL
A045125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0451500ML Supplier TCI America Supplier No. A0451500ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 693-65-2
Molecular Formula C10H22O
Molecular Weight (g/mol) 158.285
MDL Number MFCD00009501
InChI Key AOPDRZXCEAKHHW-UHFFFAOYSA-N
Synonym dipentyl ether, amyl ether, pentyl ether, n-amyl ether, diamyl ether, pentane, 1,1'-oxybis, n-pentyl ether, di-n-pentyl ether, pentyloxypentane, di-n-amyl ether
PubChem CID 12743
IUPAC Name 1-pentoxypentane
SMILES CCCCCOCCCCC

Specifications

Melting Point -69°C
Color Colorless
Boiling Point 186°C
Quantity 500 mL
UN Number 3271
Formula Weight 158.29
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Amyl Ether
EINECSNumber (2)-0365
RTECSNumber SC2900000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.