Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Amylase Monoclonal Antibody (AMY10-102.5), QED Bioscience™

Mouse Monoclonal Antibody
$250.22 - $1002.00
Specifications
Antigen | Amylase |
---|---|
Clone | AMY10-102.5 |
Concentration | 0.5-1.0 mg/mL |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-227-7525
![]() ![]() |
Leinco Technologies Inc
13501100UG |
100 μg |
Each for $250.22
|
|
|||||
50-227-7526
![]() ![]() |
Leinco Technologies Inc
13501500UG |
500 μg |
Each for $500.43
|
|
|||||
50-227-7524
![]() ![]() |
Leinco Technologies Inc
135011000UG |
1 mg |
Each for $1,002.00
|
|
|||||
Description
Amylase Monoclonal antibody specifically detects Amylase in Human samples. It is validated for ELISASpecifications
Amylase | |
0.5-1.0 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative; pH 7.4 | |
1,4-alpha-D-glucan glucanohydrolase; 1,4-alpha-D-glucan glucanohydrolase 2B; alpha-amylase; Alpha-amylase 2B; Amy2; Amy2-1; Amy2-2; Amy2a; Amy2a1; Amy2a4; AMY2B; AMY3; amylase; amylase 2-2, pancreatic; amylase 2a1; amylase 2a4; amylase 2a4, pancreatic; amylase 2b; amylase 2b, pancreatic; amylase, alpha 2A (pancreatic); amylase, alpha 2B (pancreatic); Amy-X; carcinoid alpha-amylase; ENSMUSG00000074267; glycogenase; HXA; mAmy2-1; mAmy2-3; mAmy2-6; OTTMUSG00000022432; OTTMUSG00000022433; OTTMUSG00000022462; PA; Pancreatic alpha-amylase; pancreatic amylase alpha 2A | |
Purified human pancreatic amylase. | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles |
AMY10-102.5 | |
ELISA | |
Unconjugated | |
Mouse | |
Human | |
P04746, P19961 | |
279, 280 | |
IgM | |
Protein A | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title