Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amylase Monoclonal Antibody (AMY10-105.8), QED Bioscience™
SDP

Catalog No. 502277528
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-7528 100 μg
50-227-7529 500 μg
50-227-7527 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 50-227-7528 Supplier Leinco Technologies Inc Supplier No. 13502100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Amylase Monoclonal antibody specifically detects Amylase in Human samples. It is validated for ELISA

Specifications

Antigen Amylase
Applications ELISA
Classification Monoclonal
Clone AMY10-105.8
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene AMY2A
Gene Accession No. P04746, P19961
Gene Alias 1,4-alpha-D-glucan glucanohydrolase; 1,4-alpha-D-glucan glucanohydrolase 2B; alpha-amylase; Alpha-amylase 2B; Amy2; Amy2-1; Amy2-2; Amy2a; Amy2a1; Amy2a4; AMY2B; AMY3; amylase; amylase 2-2, pancreatic; amylase 2a1; amylase 2a4; amylase 2a4, pancreatic; amylase 2b; amylase 2b, pancreatic; amylase, alpha 2A (pancreatic); amylase, alpha 2B (pancreatic); Amy-X; carcinoid alpha-amylase; ENSMUSG00000074267; glycogenase; HXA; mAmy2-1; mAmy2-3; mAmy2-6; OTTMUSG00000022432; OTTMUSG00000022433; OTTMUSG00000022462; PA; Pancreatic alpha-amylase; pancreatic amylase alpha 2A
Host Species Mouse
Immunogen Purified human pancreatic amylase.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 279, 280
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.