Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Amylin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584142
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584142 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584142 Supplier Invitrogen™ Supplier No. PA584142
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LVHSSNNFGA ILSSTNVGSN TYGKRNAVEV LKREPLNYLP Highest antigen sequence indentity to the following orthologs: Rat - 63%, Mouse - 63%.

Islet, or insulinoma, amyloid polypeptide is commonly found in pancreatic islets of patients suffering diabetes mellitus type II, or harboring an insulinoma. While the association of amylin with the development of type II diabetes has been known for some time, a direct causative role for amylin has been harder to establish. Studies suggest that amylin, like the related beta-amyloid (Abeta) associated with Alzheimer's disease, can induce apoptotic cell-death in particular cultured cells, an effect that may be relevant to the development of type II diabetes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Amylin
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IAPP
Gene Accession No. P10997
Gene Alias Amylin; amylin peptide; DAP; Diabetes-associated peptide; IAP; Iapp; Insulinoma amyloid peptide; islet amyloid polypeptide; Islet amyloid polypeptide (diabetes-associated peptide; amylin); Rabbit Anti-Amylin
Gene Symbols IAPP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human IAPP. Recombinant protein control fragment (Product #RP-101656).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3375
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.