Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Amylin Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA529713
Description
Recommended positive controls: AsPC1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Islet, or insulinoma, amyloid polypeptide is commonly found in pancreatic islets of patients suffering diabetes mellitus type II, or harboring an insulinoma. While the association of amylin with the development of type II diabetes has been known for some time, a direct causative role for amylin has been harder to establish. Studies suggest that amylin, like the related beta-amyloid (Abeta) associated with Alzheimer's disease, can induce apoptotic cell-death in particular cultured cells, an effect that may be relevant to the development of type II diabetes.
Specifications
Amylin | |
Polyclonal | |
Unconjugated | |
IAPP | |
Amylin; amylin peptide; DAP; Diabetes-associated peptide; IAP; Iapp; Insulinoma amyloid peptide; islet amyloid polypeptide; Islet amyloid polypeptide (diabetes-associated peptide; amylin); Rabbit Anti-Amylin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3375 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.83 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P10997 | |
IAPP | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human Amylin. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction