Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Amyloid A4/Amyloid β Ab-1, Mouse Monoclonal Antibody

Catalog No. MS1167S1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1167S1 500 μL Concentrated
MS1167S 1 mL Concentrated
MS1167R7 7 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MS1167S1 Supplier Epredia™ Supplier No. MS1167S1
Add to Cart
Add to Cart

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Amyloid A4/Amyloid β Ab-1, Mouse Monoclonal Antibody.

Amyloid beta-protein/amyloid A4 or amyloid beta-peptide is derived from the amyloid beta-precursor protein. Beta amyloid is an extracellular filamentous protein deposit found in the brain. It is the major protein component of amyloid cores, neuritic plaques and is also found as a deposit in neurofibrillary tangles. In humans, Alzheimer's disease is the most common cause of senile dementia and is characterized by abnormal filamentous protein deposits in the brain. Beta amyloid deposits are also detected in Lewy body dementia, Down's syndrome, amyloidosis (Dutch type) and in the Guam Parkinson-Dementia complex.

Host Species: Mouse

Clone: BAM01; same as 6F/3D

Isotype: IgG1/κ

Species Reactivity: Human. Others not tested.

Epitope: aa 8-17 (SGYEVHHQKL) of mature amyloid A4/ amyloid Beta which is the same as aa 660-669 of human amyloid A4/amyloid β protein precursor

Immunogen: A synthetic peptide consisting of residues 8-17 (SGYEVHHQKL) of the amyloid β-protein/amyloid A4 which is the same as aa 660-669 of human amyloid A4/amyloid β protein precursor

Molecular Weight: 102kDa (precursor), 4-5kDa (mature)

Positive Control: Human brain of Alzheimer's disease

Cellular Localization: Transmembrane (APP) and extracellular (Amyloid β)

Recommended for:

  • Immunohistochemistry (Formalin/paraffin only)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Amyloid A4/Amyloid beta Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone BAM01; same as 6F/3D
Conjugate Unconjugated
Host Species Mouse
Immunogen A synthetic peptide consisting of residues 8-17 (SGYEVHHQKL) of the amyloid beta-protein/amyloid A4 which is the same as aa 660-669 of human amyloid A4 / amyloid Beta protein precursor
Purification Method Human brain of Alzheimer's disease
Quantity 500 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.