Learn More
Amyloid Beta 1-16 Monoclonal Antibody (Ab5), QED Bioscience™

Mouse Monoclonal Antibody
Supplier: Leinco Technologies Inc 57004100UG

Description
This antibody recognizes an epitope within Ab1-16 as well as other Ab peptides: Ab37, Ab38, Ab39, Ab40, and Ab42. NOTE: When administered to young Tg2576 mice with minimal Ab deposition and to older mice with higher Ab loads, this antibody reduced Ab accumulation in the brain.
Amyloid Beta 1-16 Monoclonal antibody specifically detects Amyloid Beta 1-16 in Human, Mouse samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western BlotSpecifications
Amyloid Beta 1-16 | |
Monoclonal | |
0.5-1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
P05067, P12023 | |
Mouse | |
Protein A | |
RUO | |
11820, 351 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
Ab5 | |
Unconjugated | |
APP | |
A4; AAA; ABETA; Abeta40; Abeta42; ABPP; AD1; Adap; AG; AICD-50; AICD-57; AICD-59; AID(50); AID(57); AID(59); Alpha-CTF; Alpha-secretase C-terminal fragment; Alzheimer disease; Alzheimer disease amyloid A4 protein homolog; alzheimer disease amyloid protein; Amyloid; amyloid A4; Amyloid b; Amyloid beta; amyloid beta (A4) precursor protein; amyloid beta (A4) precursor protein (peptidase nexin-II, Alzheimer disease); amyloid beta A4 protein; amyloid beta precursor protein; Amyloid intracellular domain 50; Amyloid intracellular domain 57; Amyloid intracellular domain 59; Amyloid precursor protein; amyloid precursor protein variant 1; amyloid precursor protein variant 2; Amyloid β; amyloid-beta A4 protein; amyloid-beta A4 protein; amyloid beta A4 protein; Amyloid-beta precursor protein; Amyloid-beta protein 40; Amyloid-beta protein 42; Amyloidogenic glycoprotein; APP; APP-C57; APP-C59; APP-C99; APPI; appican; beta amyloid protein; beta amyloid protein precursor; beta-amyloid peptide; beta-amyloid peptide(1-40); beta-amyloid peptide(1-42); Beta-amyloid precursor protein; betaApp; Beta-APP40; Beta-APP42; Beta-CTF; Beta-secretase C-terminal fragment; C31; C80; C83; C99; Cerebral vascular amyloid peptide; CTF gamma; CTF-alpha; CTFgamma; CVAP; E030013M08Rik; Gamma-CTF(50); Gamma-CTF(57); Gamma-CTF(59); Gamma-secretase C-terminal fragment 50; Gamma-secretase C-terminal fragment 57; Gamma-secretase C-terminal fragment 59; N-APP; OTTHUMP00000096096; P3(40); P3(42); Pan-Abeta; peptidase nexin-II; PN2; PN-II; PreA4; PreA4 751; protease nexin II; protease nexin-II; S-APP-alpha; S-APP-beta; Soluble APP-alpha; Soluble APP-beta; testicular tissue protein Li 2 | |
Aggregated Amyloid beta 1-42, fibrillar Amyloid beta . | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.