Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amyloid beta fibrils Rabbit anti-Human, Mouse, Clone: M98, MilliporeSigma™

Catalog No. MABN640I25 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN640I25 25 μg
MABN640I100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABN640I25 Supplier MilliporeSigma Supplier No. MABN640I25UG
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Amyloid beta fibrils Monoclonal antibody specifically detects Amyloid beta fibrils in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Amyloid beta fibrils
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone M98
Conjugate Unconjugated
Formulation Purified rabbit recombinant monoclonal antibody in PBS with 0.05% sodium azide.
Host Species Rabbit
Immunogen Fibrillar Abeta 1-42 peptide
Purification Method Protein A purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.