Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AMZ1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550605
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550605 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550605 Supplier Invitrogen™ Supplier No. PA550605
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total AMZ1 protein.

Zinc metalloprotease. Exhibits aminopeptidase activity against neurogranin in vitro. Does not hydrolyze angiotensin-2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AMZ1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Amz1
Gene Accession No. Q400G9
Gene Alias 5330426I05; 6530401C20Rik; AMZ1; archaelysin family metallopeptidase 1; archaemetzincin-1; archeobacterial metalloproteinase-like 1; archeobacterial metalloproteinase-like protein 1; KIAA1950; metalloproteinase-like protein; RGD1311314
Gene Symbols Amz1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues near the C terminal of human archaelysin family metallopeptidase 1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 155185
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.