Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANKRD13A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109390
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109390 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109390 Supplier Invitrogen™ Supplier No. PA5109390
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSSACDAGDH YPLHLLVWKN DYRQLEKELQ GQNVEAVDPR GRTLLHLAVS LGHLESARVL LRHKADVTKE NRQGWTVLHE AVSTGDPEMV YTVLQHRDYH NTSMALEGVP ELLQKILEAP DFYVQMKWEF TSWVPLVSRI CPNDVCRIWK SGAKLRVDIT LLGFENMSWI RGRRSFIFKG EDNWAELMEV NHDDKVVTTE RFDLSQEMER LTLDLMKPKS REVERRLTSP VINTSLDTKN.

ANKRD13A, also named as ANKRD13, a 590 amino acid protein, which contains two ANK repeats and four UIM (ubiquitin-interacting motif) repeats. ANKRD13A localizes in the cell membrane. ANKRD13A as ubiquitin-binding protein that specifically recognizes and binds 'Lys-63'-linked ubiquitin, not bind 'Lys-48'-linked ubiquitin. ANKRD13A positively regulates the internalization of ligand-activated EGFR by binding to the Ub moiety of ubiquitinated EGFR at the cell membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKRD13A
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.63 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ANKRD13A
Gene Accession No. Q8IZ07
Gene Alias 1100001D10Rik; Ankrd13; ANKRD13A; ankyrin repeat domain 13; ankyrin repeat domain 13A; ankyrin repeat domain-containing protein 13A; AU046136; NY-REN-25; NY-REN-25 antigen; Protein KE03
Gene Symbols ANKRD13A
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human ANKRD13A (NP_149112.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 88455
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.