Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AN13D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570180
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570180 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570180 Supplier Invitrogen™ Supplier No. PA570180
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 79%; Human: 100%; Mouse: 90%; Rabbit: 86%; Rat: 90%.

The protein encoded by this gene is a member of the ankyrin repeat domain (ANKRD) 13 family, which currently consists of four proteins containing ubiquitin-interacting motifs. These proteins are integral membrane proteins that bind specifically to Lys-63-linked ubiquitin chains on membrane-bound proteins, targeting those proteins for rapid internalization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AN13D
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ANKRD13D
Gene Accession No. Q6ZTN6
Gene Alias 0710001P18Rik; AI430801; ANKRD13D; ankyrin repeat domain 13 family member D; ankyrin repeat domain 13 family, member D; ankyrin repeat domain 13D; ankyrin repeat domain-containing protein 13D; RGD1309156
Gene Symbols ANKRD13D
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human AN13D.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 338692
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.